siRNA Details
Gene Symbol: TBC1D7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143964.3 | NP_001137436.1 | 4 | 506 | ||
NM_001143965.3 | NP_001137437.1 | 4 | 366 | |||
NM_001143966.3 | NP_001137438.1 | 3 | 285 | |||
NM_001258457.2 | NP_001245386.1 | 4 | 366 | |||
NM_001318805.1 | NP_001305734.1 | 4 | 423 | |||
NM_001318806.1 | NP_001305735.1 | 3 | 285 | |||
NM_016495.5 | NP_057579.1 | 4 | 526 | |||
GenBank mRNA | AB449888.1 | - | 3 | 230 | ||
AF151073.1 | - | 4 | 323 | |||
AK223445.1 | - | 4 | 403 | |||
AL833474.1 | - | 3 | 258 | |||
AY542308.1 | - | 4 | 288 | |||
AY826820.1 | - | 3 | 211 | |||
BC007054.1 | - | 4 | 327 | 3828592 | ||
BC050465.1 | - | 3 | 227 | 6153876 | ||
BG504676.1 | - | 4 | 539 | |||
BI914228.1 | - | 4 | 487 | |||
BP319580.1 | - | 4 | 297 | |||
BX354770.2 | - | 4 | 363 | |||
DA169008.1 | - | 3 | 380 |
Gene Symbol: TBC1D7-LOC100130357 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318809.1 | NP_001305738.1 | 4 | 366 | ||
NR_134872.1 | 4 | 366 | ||||
GenBank mRNA | AK057228.1 | - | 4 | 326 | ||
AK296391.1 | - | 4 | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
