siRNA Details
Gene Symbol: TBC1D7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143964.3 | NP_001137436.1 | 7 | 1053 | ||
NM_001143965.3 | NP_001137437.1 | 7 | 913 | |||
NM_001143966.3 | NP_001137438.1 | 6 | 832 | |||
NM_001258457.2 | NP_001245386.1 | 6 | 775 | |||
NM_001318805.1 | NP_001305734.1 | 7 | 970 | |||
NM_016495.5 | NP_057579.1 | 7 | 1073 | |||
GenBank mRNA | AB449888.1 | - | 6 | 777 | ||
AF151073.1 | - | 7 | 870 | |||
AK223445.1 | - | 7 | 950 | |||
AY542308.1 | - | 6 | 697 | |||
AY826820.1 | - | 6 | 758 | |||
BC007054.1 | - | 7 | 874 | 3828592 | ||
BC050465.1 | - | 6 | 774 | 6153876 |
Gene Symbol: TBC1D7-LOC100130357 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318809.1 | NP_001305738.1 | 7 | 913 | ||
GenBank mRNA | AK057228.1 | - | 7 | 873 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
