siRNA Details
Gene Symbol: OR4A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005272.3 | NP_001005272.3 | 1 | 257 | ||
GenBank mRNA | BC140750.1 | - | 1 | 257 | 9021618 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
