siRNA Details
Gene Symbol: GPAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321525.1 | NP_001308454.1 | 18 | 2423 | ||
NM_001321526.1 | NP_001308455.1 | 18 | 2513 | |||
NM_001321527.1 | NP_001308456.1 | 18 | 2477 | |||
NM_001321528.1 | NP_001308457.1 | 18 | 2477 | |||
NM_001321529.1 | NP_001308458.1 | 17 | 2300 | |||
NM_001321530.1 | NP_001308459.1 | 17 | 2192 | |||
NM_207328.3 | NP_997211.2 | 18 | 2495 | |||
XM_006712308.3 | XP_006712371.1 | 18 | 2188 | |||
XM_006712311.3 | XP_006712374.1 | 18 | 2188 | |||
XM_017003424.1 | XP_016858913.1 | 18 | 2127 | |||
GenBank mRNA | AK122580.1 | - | 9 | 3837 | ||
AK122734.1 | - | 10 | 1235 | |||
AK131197.1 | - | 12 | 1532 | |||
AK298131.1 | - | 17 | 1872 | |||
AK302887.1 | - | 16 | 3227 | |||
BC068596.1 | - | 18 | 2166 | 4837765 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
