siRNA Details
Gene Symbol: LHFPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199000.2 | NP_945351.1 | 1 | 309 | ||
XM_005250327.1 | XP_005250384.1 | 1 | 186 | |||
GenBank mRNA | AK124251.1 | - | 1 | 78 | ||
AK299759.1 | - | 1 | 188 | |||
AY260763.1 | - | 1 | 309 | |||
BC129937.1 | - | 1 | 265 | 40116045 | ||
BC129938.1 | - | 1 | 265 | 40116047 | ||
BC129941.1 | - | 1 | 265 | 40118480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
