siRNA Details
Gene Symbol: DNAJB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039550.1 | NP_001034639.1 | 3 | 332 | ||
NM_006736.5 | NP_006727.2 | 3 | 332 | |||
GenBank mRNA | AK289626.1 | - | 3 | 155 | ||
AK292761.1 | - | 3 | 160 | |||
AK293888.1 | - | 3 | 284 | |||
AK312723.1 | - | 3 | 339 | |||
AL547521.3 | - | 3 | 378 | |||
BC011609.2 | - | 3 | 274 | 3619734 | ||
BC047056.1 | - | 2 | 135 | 5185009 | ||
BT007088.1 | - | 2 | 95 | |||
BX420085.2 | - | 334 | ||||
S37374.1 | - | 2 | 120 | |||
S37375.1 | - | 2 | 120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
