siRNA Details
Gene Symbol: AGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037131.2 | NP_001032208.1 | 16 | 2551 | ||
NM_014914.4 | NP_055729.2 | 15 | 2392 | |||
XM_005246059.4 | XP_005246116.1 | 16 | 1857 | |||
XM_006712234.3 | XP_006712297.1 | 18 | 2092 | |||
XM_006712235.3 | XP_006712298.1 | 17 | 2216 | |||
XM_006712237.3 | XP_006712300.2 | 17 | 2873 | |||
XM_006712239.3 | XP_006712302.2 | 18 | 2888 | |||
XM_011510547.2 | XP_011508849.1 | 17 | 2809 | |||
XM_011510548.2 | XP_011508850.1 | 17 | 2727 | |||
XM_011510549.2 | XP_011508851.1 | 16 | 2690 | |||
XM_017003282.1 | XP_016858771.1 | 17 | 2762 | |||
GenBank mRNA | AB029022.1 | - | 15 | 2365 | ||
AF413078.1 | - | 15 | 1794 | |||
AK001722.1 | - | 6 | 671 | |||
AK295520.1 | - | 4 | 368 | |||
AK300763.1 | - | 12 | 1491 | |||
AY033765.1 | - | 15 | 2355 | |||
BC140856.1 | - | 17 | 2168 | 9021724 | ||
BC167153.1 | - | 15 | 2365 | 8862550 | ||
BC167157.1 | - | 15 | 2377 | 8862571 | ||
BI868924.1 | - | 2 | 109 | |||
KJ534764.1 | - | 16 | 1806 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
