siRNA Details
Gene Symbol: AGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037131.2 | NP_001032208.1 | 15 | 2450 | ||
NM_014914.4 | NP_055729.2 | 14 | 2291 | |||
XM_005246059.4 | XP_005246116.1 | 15 | 1756 | |||
XM_006712234.3 | XP_006712297.1 | 17 | 1991 | |||
XM_006712235.3 | XP_006712298.1 | 16 | 2115 | |||
XM_006712237.3 | XP_006712300.2 | 16 | 2772 | |||
XM_006712239.3 | XP_006712302.2 | 17 | 2787 | |||
XM_011510547.2 | XP_011508849.1 | 16 | 2708 | |||
XM_011510548.2 | XP_011508850.1 | 16 | 2626 | |||
XM_011510549.2 | XP_011508851.1 | 15 | 2589 | |||
XM_017003282.1 | XP_016858771.1 | 16 | 2661 | |||
GenBank mRNA | AB029022.1 | - | 14 | 2264 | ||
AF413078.1 | - | 14 | 1693 | |||
AK001722.1 | - | 5 | 570 | |||
AK295520.1 | - | 3 | 267 | |||
AK300763.1 | - | 11 | 1390 | |||
AY033765.1 | - | 14 | 2254 | |||
BC140856.1 | - | 16 | 2067 | 9021724 | ||
BC167153.1 | - | 14 | 2264 | 8862550 | ||
BC167157.1 | - | 14 | 2276 | 8862571 | ||
KJ534764.1 | - | 15 | 1705 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
