siRNA Details
Gene Symbol: CABIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199281.1 | NP_001186210.1 | 36, 37 | 6947 | ||
NM_001201429.1 | NP_001188358.1 | 35, 36 | 6491 | |||
NM_012295.3 | NP_036427.1 | 36, 37 | 6641 | |||
XM_005261415.2 | XP_005261472.1 | 35, 36 | 6693 | |||
XM_005261416.2 | XP_005261473.1 | 35, 36 | 6678 | |||
XM_005261417.2 | XP_005261474.1 | 35, 36 | 6603 | |||
XM_005261419.2 | XP_005261476.1 | 35, 36 | 6734 | |||
XM_006724179.1 | XP_006724242.1 | 35, 36 | 6606 | |||
XM_006724180.1 | XP_006724243.1 | 35, 36 | 6516 | |||
XM_006724181.1 | XP_006724244.1 | 35, 36 | 6501 | |||
XM_011530026.1 | XP_011528328.1 | 34, 35 | 6498 | |||
XM_017028680.1 | XP_016884169.1 | 35, 36 | 6591 | |||
XM_017028681.1 | XP_016884170.1 | 34, 35 | 6543 | |||
XM_017028682.1 | XP_016884171.1 | 35, 36 | 6614 | |||
XM_017028683.1 | XP_016884172.1 | 34, 35 | 6351 | |||
XM_017028684.1 | XP_016884173.1 | 20, 21 | 4231 | |||
GenBank mRNA | AB002328.2 | - | 36, 37 | 6864 | ||
AF072441.1 | - | 36, 37 | 6642 | |||
AL390166.1 | - | 7, 8 | 1673 | |||
BC021929.1 | - | 11, 12 | 2421 | |||
BC054497.1 | - | 35, 36 | 6371 | 4521929 |

More Information
Other Species Targeted

Gene Ontology Categories:
