siRNA Details
Gene Symbol: VSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288791.1 | NP_001275720.1 | 9 | 654 | ||
NM_001288792.1 | NP_001275721.1 | 8 | 770 | |||
NM_001288793.1 | NP_001275722.1 | 6 | 503 | |||
NM_198481.3 | NP_940883.2 | 9 | 863 | |||
NR_110142.1 | 10 | 952 | ||||
XM_011526845.1 | XP_011525147.1 | 10 | 1260 | |||
XM_011526846.1 | XP_011525148.1 | 10 | 1170 | |||
XM_011526847.1 | XP_011525149.1 | 11 | 1238 | |||
XM_011526848.1 | XP_011525150.1 | 9 | 1077 | |||
XM_011526849.1 | XP_011525151.1 | 9 | 966 | |||
GenBank mRNA | AY358542.1 | - | 9 | 872 | ||
BC100943.2 | - | 8 | 747 | 40004095 | ||
BG189223.1 | - | 5 | 302 | |||
DQ479397.1 | - | 9 | 704 | |||
FJ584316.1 | - | 9 | 698 | |||
FJ584317.1 | - | 9 | 698 | |||
FJ584318.1 | - | 8 | 605 | |||
FJ584319.1 | - | 6 | 338 | |||
FJ584320.1 | - | 9 | 489 | |||
FJ882051.1 | - | 10 | 787 | |||
FN398145.1 | - | 9 | 691 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
