siRNA Details
Gene Symbol: DRAXIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198545.3 | NP_940947.3 | 4, 5 | 896 | ||
XM_011541405.2 | XP_011539707.1 | 5, 6 | 1070 | |||
XM_017001214.1 | XP_016856703.1 | 5, 6 | 1206 | |||
GenBank mRNA | AB301919.1 | - | 3, 4 | 756 | ||
AK075558.1 | - | 4, 5 | 891 | |||
AK311439.1 | - | 1, 2 | 44 | |||
AY358750.1 | - | 4, 5 | 857 | |||
BC021286.2 | - | 2, 3 | 246 | |||
BC111064.1 | - | 4, 5 | 898 | 5769984 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
