siRNA Details
Gene Symbol: DRAXIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198545.3 | NP_940947.3 | 6 | 1043 | ||
XM_011541405.2 | XP_011539707.1 | 7 | 1217 | |||
XM_017001214.1 | XP_016856703.1 | 7 | 1353 | |||
GenBank mRNA | AB301919.1 | - | 5 | 903 | ||
AK075558.1 | - | 6 | 1038 | |||
AK311439.1 | - | 3 | 191 | |||
AY358750.1 | - | 6 | 1004 | |||
BC021286.2 | - | 4 | 393 | |||
BC111064.1 | - | 6 | 1045 | 5769984 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
