siRNA Details
Gene Symbol: DRAXIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198545.3 | NP_940947.3 | 3 | 655 | ||
XM_011541405.2 | XP_011539707.1 | 4 | 829 | |||
XM_017001214.1 | XP_016856703.1 | 4 | 965 | |||
GenBank mRNA | AB301919.1 | - | 2 | 515 | ||
AK075558.1 | - | 3 | 650 | |||
AY358750.1 | - | 3 | 616 | |||
BC111064.1 | - | 3 | 657 | 5769984 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
