siRNA Details
Gene Symbol: ACIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164814.1 | NP_001158286.1 | 15 | 3431 | ||
NM_001164815.1 | NP_001158287.1 | 14 | 3350 | |||
NM_001164816.1 | NP_001158288.1 | 8 | 989 | |||
NM_001164817.1 | NP_001158289.1 | 9 | 1051 | |||
NM_014977.3 | NP_055792.1 | 15 | 3470 | |||
XM_005267415.3 | XP_005267472.1 | 15 | 3271 | |||
XM_005267416.3 | XP_005267473.1 | 15 | 3238 | |||
XM_005267418.1 | XP_005267475.1 | 8 | 1286 | |||
GenBank mRNA | AB014570.2 | - | 15 | 2979 | ||
AF124726.1 | - | 15 | 3469 | |||
AF124727.1 | - | 9 | 1005 | |||
AF124728.1 | - | 8 | 980 | |||
AK299034.1 | - | 14 | 2861 | |||
AL050382.1 | - | 8 | 2075 | |||
AL833228.1 | - | 10 | 1320 | |||
BC140805.1 | - | 15 | 3200 | 9021673 | ||
BX247975.1 | - | 8 | 1259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
