siRNA Details
Gene Symbol: ACIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164814.1 | NP_001158286.1 | 9 | 2663 | ||
NM_001164815.1 | NP_001158287.1 | 8 | 2543 | |||
NM_001164816.1 | NP_001158288.1 | 2 | 182 | |||
NM_001164817.1 | NP_001158289.1 | 3 | 244 | |||
NM_014977.3 | NP_055792.1 | 9 | 2663 | |||
XM_005267415.3 | XP_005267472.1 | 9 | 2467 | |||
XM_005267416.3 | XP_005267473.1 | 9 | 2467 | |||
XM_005267418.1 | XP_005267475.1 | 2 | 479 | |||
XM_006720081.3 | XP_006720144.1 | 9 | 2467 | |||
XM_011536570.2 | XP_011534872.1 | 9 | 2467 | |||
GenBank mRNA | AB014570.2 | - | 9 | 2172 | ||
AF124726.1 | - | 9 | 2662 | |||
AF124727.1 | - | 3 | 198 | |||
AF124728.1 | - | 2 | 173 | |||
AK299034.1 | - | 8 | 2054 | |||
AL833228.1 | - | 4 | 516 | |||
BC140805.1 | - | 9 | 2432 | 9021673 | ||
BX247975.1 | - | 2 | 452 | |||
CX786860.1 | - | 3 | 212 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
