siRNA Details
Gene Symbol: ACIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164814.1 | NP_001158286.1 | 11, 12 | 2952 | ||
NM_001164815.1 | NP_001158287.1 | 11 | 2871 | |||
NM_001164816.1 | NP_001158288.1 | 5 | 510 | |||
NM_001164817.1 | NP_001158289.1 | 6 | 572 | |||
NM_014977.3 | NP_055792.1 | 12 | 2991 | |||
XM_005267415.3 | XP_005267472.1 | 12 | 2792 | |||
XM_005267416.3 | XP_005267473.1 | 11, 12 | 2759 | |||
XM_005267418.1 | XP_005267475.1 | 5 | 807 | |||
GenBank mRNA | AB014570.2 | - | 12 | 2500 | ||
AF124726.1 | - | 12 | 2990 | |||
AF124727.1 | - | 6 | 526 | |||
AF124728.1 | - | 5 | 501 | |||
AK299034.1 | - | 11 | 2382 | |||
AL050382.1 | - | 4, 5 | 1596 | |||
AL833228.1 | - | 7 | 841 | |||
BC140805.1 | - | 11, 12 | 2721 | 9021673 | ||
BX247975.1 | - | 5 | 780 | |||
CX786860.1 | - | 5, 6 | 504 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
