siRNA Details
Gene Symbol: ZNF81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007137.3 | NP_009068.2 | 5 | 988 | ||
XM_005272600.3 | XP_005272657.1 | 5 | 1046 | |||
XM_011543899.2 | XP_011542201.1 | 5 | 2652 | |||
XM_011543900.2 | XP_011542202.1 | 6 | 1107 | |||
XM_017029486.1 | XP_016884975.1 | 6 | 1165 | |||
XM_017029487.1 | XP_016884976.1 | 5 | 1398 | |||
GenBank mRNA | AY487248.1 | - | 4 | 739 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
