siRNA Details
Gene Symbol: PHF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184896.1 | NP_001171825.1 | 20 | 2953 | ||
NM_001184897.1 | NP_001171826.1 | 19 | 2801 | |||
NM_001184898.1 | NP_001171827.1 | 21 | 2724 | |||
NM_015107.2 | NP_055922.1 | 20 | 3104 | |||
XM_005261996.1 | XP_005262053.1 | 21 | 2797 | |||
XM_005261997.3 | XP_005262054.1 | 20 | 3402 | |||
XM_005261999.1 | XP_005262056.1 | 20 | 2773 | |||
XM_005262000.1 | XP_005262057.1 | 19 | 2763 | |||
XM_011530778.1 | XP_011529080.1 | 20 | 3066 | |||
XM_017029361.1 | XP_016884850.1 | 20 | 3352 | |||
XM_017029362.1 | XP_016884851.1 | 20 | 3352 | |||
GenBank mRNA | AB029034.1 | - | 20 | 2780 | ||
AF091081.1 | - | 14 | 1895 | |||
AK021696.1 | - | 18 | 2305 | |||
AK022788.1 | - | 8 | 1272 | |||
AK304272.1 | - | 19 | 2623 | |||
BC053861.1 | - | 20 | 2808 | 5138930 | ||
CR933612.1 | - | 20 | 2939 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
