siRNA Details
Gene Symbol: PHF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184896.1 | NP_001171825.1 | 17 | 2441 | ||
NM_001184897.1 | NP_001171826.1 | 16 | 2289 | |||
NM_015107.2 | NP_055922.1 | 17 | 2592 | |||
XM_005261996.1 | XP_005262053.1 | 18 | 2285 | |||
XM_005261997.3 | XP_005262054.1 | 17 | 2890 | |||
XM_005261999.1 | XP_005262056.1 | 17 | 2261 | |||
XM_005262000.1 | XP_005262057.1 | 16 | 2251 | |||
XM_011530778.1 | XP_011529080.1 | 17 | 2554 | |||
XM_017029361.1 | XP_016884850.1 | 17 | 2840 | |||
XM_017029362.1 | XP_016884851.1 | 17 | 2840 | |||
GenBank mRNA | AB029034.1 | - | 17 | 2268 | ||
AF091081.1 | - | 11 | 1383 | |||
AK021696.1 | - | 15 | 1793 | |||
AK022788.1 | - | 5 | 760 | |||
AK304272.1 | - | 16 | 2111 | |||
BC053861.1 | - | 17 | 2296 | 5138930 | ||
CR933612.1 | - | 17 | 2427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
