siRNA Details
Gene Symbol: FBXL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152441.2 | NP_689654.1 | 1 | 113 | ||
XM_017018875.1 | XP_016874364.1 | 1 | 137 | |||
XM_017018876.1 | XP_016874365.1 | 1 | 137 | |||
GenBank mRNA | BC028132.1 | - | 1 | 113 | 5205284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
