siRNA Details
Gene Symbol: NBEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204197.1 | NP_001191126.1 | 11 | 1287 | ||
NM_015678.4 | NP_056493.3 | 51 | 8317 | |||
XM_005266346.4 | XP_005266403.1 | 51 | 8302 | |||
XM_005266347.4 | XP_005266404.2 | 52 | 8380 | |||
XM_005266348.4 | XP_005266405.2 | 51 | 8371 | |||
XM_006719803.3 | XP_006719866.1 | 52 | 8365 | |||
XM_006719805.3 | XP_006719868.1 | 50 | 8293 | |||
XM_006719806.3 | XP_006719869.1 | 50 | 8308 | |||
XM_011535046.2 | XP_011533348.1 | 44 | 6511 | |||
XM_017020544.1 | XP_016876033.1 | 52 | 8362 | |||
XM_017020545.1 | XP_016876034.1 | 51 | 8299 | |||
XM_017020546.1 | XP_016876035.1 | 52 | 7668 | |||
GenBank mRNA | AB046764.1 | - | 18 | 2029 | ||
AF072371.1 | - | 5 | 504 | |||
AF467288.1 | - | 52 | 7989 | |||
AK091469.1 | - | 22 | 2476 | |||
AK122888.1 | - | 21 | 2830 | |||
AK127791.1 | - | 28 | 3668 | |||
AK294737.1 | - | 11 | 1287 | |||
AK297773.1 | - | 12 | 1385 | |||
AL137748.1 | - | 6 | 992 | |||
BX640760.1 | - | 27 | 3647 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
