siRNA Details
Gene Symbol: NBEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204197.1 | NP_001191126.1 | 12 | 1464 | ||
NM_015678.4 | NP_056493.3 | 52 | 8494 | |||
XM_005266346.4 | XP_005266403.1 | 52 | 8479 | |||
XM_005266347.4 | XP_005266404.2 | 53 | 8557 | |||
XM_005266348.4 | XP_005266405.2 | 52 | 8548 | |||
XM_006719803.3 | XP_006719866.1 | 53 | 8542 | |||
XM_006719805.3 | XP_006719868.1 | 51 | 8470 | |||
XM_006719806.3 | XP_006719869.1 | 51 | 8485 | |||
XM_011535046.2 | XP_011533348.1 | 45 | 6688 | |||
XM_017020544.1 | XP_016876033.1 | 53 | 8539 | |||
XM_017020545.1 | XP_016876034.1 | 52 | 8476 | |||
XM_017020546.1 | XP_016876035.1 | 53 | 7845 | |||
GenBank mRNA | AB046764.1 | - | 19 | 2206 | ||
AF072371.1 | - | 6 | 681 | |||
AF467288.1 | - | 53 | 8166 | |||
AK091469.1 | - | 23 | 2653 | |||
AK122888.1 | - | 22 | 3007 | |||
AK127791.1 | - | 29 | 3845 | |||
AK294737.1 | - | 12 | 1464 | |||
AL137748.1 | - | 7 | 1169 | |||
BX640760.1 | - | 28 | 3824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
