siRNA Details
Gene Symbol: NBEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204197.1 | NP_001191126.1 | 13 | 1488 | ||
NM_015678.4 | NP_056493.3 | 53 | 8518 | |||
XM_005266346.4 | XP_005266403.1 | 53 | 8503 | |||
XM_005266347.4 | XP_005266404.2 | 54 | 8581 | |||
XM_005266348.4 | XP_005266405.2 | 53 | 8572 | |||
XM_006719803.3 | XP_006719866.1 | 54 | 8566 | |||
XM_006719805.3 | XP_006719868.1 | 52 | 8494 | |||
XM_006719806.3 | XP_006719869.1 | 52 | 8509 | |||
XM_011535046.2 | XP_011533348.1 | 46 | 6712 | |||
XM_017020544.1 | XP_016876033.1 | 54 | 8563 | |||
XM_017020545.1 | XP_016876034.1 | 53 | 8500 | |||
XM_017020546.1 | XP_016876035.1 | 54 | 7869 | |||
GenBank mRNA | AB046764.1 | - | 20 | 2230 | ||
AF072371.1 | - | 7 | 705 | |||
AF467288.1 | - | 54 | 8190 | |||
AK091469.1 | - | 24 | 2677 | |||
AK122888.1 | - | 23 | 3031 | |||
AK127791.1 | - | 30 | 3869 | |||
AK294737.1 | - | 13 | 1488 | |||
AL137748.1 | - | 8 | 1193 | |||
BX640760.1 | - | 29 | 3848 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
