siRNA Details
Gene Symbol: TMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080532.1 | NP_001074001.1 | 14, 15 | 1640 | ||
GenBank mRNA | AY263163.1 | - | 14, 15 | 1679 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
