siRNA Details
Gene Symbol: TMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080532.1 | NP_001074001.1 | 11 | 1120 | ||
GenBank mRNA | AY236490.1 | - | 11 | 1198 | ||
AY263163.1 | - | 11 | 1159 | |||
BC132679.1 | - | 11 | 1157 | 40146701 | ||
BC143410.1 | - | 11 | 1157 | 9051917 | ||
BC157874.1 | - | 11 | 1169 | 9057092 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
