siRNA Details
Gene Symbol: CMPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256477.1 | NP_001243406.1 | 3 | 937 | ||
NM_001256478.1 | NP_001243407.1 | 3 | 937 | |||
NM_207315.3 | NP_997198.2 | 3 | 937 | |||
NR_046236.1 | 3 | 349 | ||||
GenBank mRNA | AK127983.1 | - | 3 | 1025 | ||
AK308509.1 | - | 4 | 461 | |||
BC089425.1 | - | 3 | 790 | |||
BC132821.1 | - | 3 | 937 | 40146843 | ||
BC141802.1 | - | 3 | 830 | 40147165 | ||
BC144202.1 | - | 3 | 937 | 9052722 | ||
BX463706.2 | - | 3 | 245 | |||
DA501203.1 | - | 3 | 246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
