siRNA Details
Gene Symbol: DNAJB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153614.3 | NP_705842.2 | 7 | 1511 | ||
XM_005273984.3 | XP_005274041.1 | 6 | 876 | |||
XM_011545004.2 | XP_011543306.1 | 9 | 1762 | |||
XM_011545005.2 | XP_011543307.1 | 8 | 1602 | |||
XM_011545007.2 | XP_011543309.1 | 7 | 1287 | |||
XM_011545009.2 | XP_011543311.1 | 9 | 1436 | |||
XM_011545015.2 | XP_011543317.1 | 4 | 629 | |||
XM_017017675.1 | XP_016873164.1 | 8 | 1032 | |||
XM_017017676.1 | XP_016873165.1 | 9 | 1612 | |||
XM_017017677.1 | XP_016873166.1 | 8 | 1481 | |||
XM_017017678.1 | XP_016873167.1 | 7 | 1292 | |||
GenBank mRNA | AF516185.1 | - | 4 | 1483 | ||
AK058095.1 | - | 4 | 1483 | |||
AK128589.1 | - | 9 | 3556 | |||
AY138810.2 | - | 7 | 1511 | |||
AY325766.1 | - | 7 | 804 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
