siRNA Details
Gene Symbol: DCDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020869.3 | NP_065920.2 | 11 | 1675 | ||
GenBank mRNA | AB040926.1 | - | 1 | 201 | ||
BX647348.1 | - | 11 | 1675 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
