siRNA Details
Gene Symbol: SP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080391.1 | NP_001073860.1 | 3 | 389 | ||
NM_001206701.1 | NP_001193630.1 | 3 | 389 | |||
NM_001206702.1 | NP_001193631.1 | 3 | 389 | |||
NM_001206703.1 | NP_001193632.1 | 2 | 314 | |||
NM_001206704.1 | NP_001193633.1 | 4 | 424 | |||
NM_003113.3 | NP_003104.2 | 3 | 389 | |||
GenBank mRNA | AF056322.1 | - | 3 | 279 | ||
AF255565.1 | - | 3 | 279 | |||
AK091898.1 | - | 4 | 424 | |||
AK293373.1 | - | 2 | 251 | |||
AK304205.1 | - | 3 | 346 | |||
BC011562.1 | - | 3 | 327 | 4560914 | ||
M60618.1 | - | 3 | 279 | |||
U36501.1 | - | 3 | 279 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
