siRNA Details
Gene Symbol: CD28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243077.1 | NP_001230006.1 | 1 | 256 | ||
NM_001243078.1 | NP_001230007.1 | 1 | 256 | |||
NM_006139.3 | NP_006130.1 | 1 | 256 | |||
XM_011512197.2 | XP_011510499.1 | 1 | 107 | |||
GenBank mRNA | AF222341.1 | - | 1 | 34 | ||
AF222342.1 | - | 1 | 34 | |||
AF222343.1 | - | 1 | 34 | |||
AJ295273.1 | - | 1 | 34 | |||
AK292986.1 | - | 1 | 38 | |||
AK303421.1 | - | 1 | 93 | |||
AK313313.1 | - | 1 | 180 | |||
BC093698.1 | - | 1 | 74 | 7939543 | ||
BC112085.1 | - | 1 | 74 | 8327553 | ||
J02988.1 | - | 1 | 133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
