siRNA Details
Gene Symbol: CD28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243077.1 | NP_001230006.1 | 3 | 353 | ||
NM_001243078.1 | NP_001230007.1 | 2 | 287 | |||
NM_006139.3 | NP_006130.1 | 3 | 644 | |||
XM_011512194.2 | XP_011510496.1 | 3 | 479 | |||
XM_011512195.2 | XP_011510497.1 | 4 | 221 | |||
XM_011512197.2 | XP_011510499.1 | 4 | 243 | |||
GenBank mRNA | AF222341.1 | - | 3 | 131 | ||
AF222342.1 | - | 2 | 65 | |||
AF222343.1 | - | 4 | 170 | |||
AJ517504.1 | - | 3 | 464 | |||
AK292986.1 | - | 3 | 426 | |||
AK303421.1 | - | 4 | 404 | |||
AK313313.1 | - | 3 | 568 | |||
BC093698.1 | - | 3 | 462 | 7939543 | ||
BC112085.1 | - | 3 | 462 | 8327553 | ||
J02988.1 | - | 3 | 521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
