siRNA Details
Gene Symbol: BRCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018055.2 | NP_001018065.1 | 4 | 317 | ||
NM_001242640.1 | NP_001229569.1 | 4 | 320 | |||
NM_024332.3 | NP_077308.1 | 4 | 317 | |||
XM_005274751.4 | XP_005274808.1 | 4 | 330 | |||
XM_017029838.1 | XP_016885327.1 | 4 | 334 | |||
GenBank mRNA | AK298886.1 | - | 3 | 152 | ||
AK299194.1 | - | 4 | 215 | |||
AK310286.1 | - | 4 | 218 | |||
AK313544.1 | - | 4 | 212 | |||
AY438030.1 | - | 4 | 209 | |||
BC002999.1 | - | 4 | 211 | 2821917 | ||
BC006540.2 | - | 4 | 211 | 2821917 | ||
BP304352.1 | - | 4 | 320 | |||
BU902387.1 | - | 349 | ||||
S68015.1 | - | 4 | 242 | |||
X64643.1 | - | 4 | 215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
