siRNA Details
Gene Symbol: BRCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018055.2 | NP_001018065.1 | 9, 10 | 830 | ||
NM_001242640.1 | NP_001229569.1 | 9, 10 | 833 | |||
NM_024332.3 | NP_077308.1 | 10, 11 | 905 | |||
XM_005274751.4 | XP_005274808.1 | 10, 11 | 918 | |||
XM_017029838.1 | XP_016885327.1 | 9, 10 | 847 | |||
GenBank mRNA | AK298886.1 | - | 9, 10 | 740 | ||
AK299194.1 | - | 9, 10 | 728 | |||
AK313544.1 | - | 10, 11 | 800 | |||
AY438030.1 | - | 10, 11 | 797 | |||
BC002999.1 | - | 10, 11 | 799 | 2821917 | ||
BC006540.2 | - | 10, 11 | 799 | 2821917 | ||
S68015.1 | - | 10, 11 | 830 | |||
X64643.1 | - | 9, 10 | 728 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
