siRNA Details
Gene Symbol: BRCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018055.2 | NP_001018065.1 | 7 | 621 | ||
NM_001242640.1 | NP_001229569.1 | 7 | 624 | |||
NM_024332.3 | NP_077308.1 | 7 | 621 | |||
XM_005274751.4 | XP_005274808.1 | 7 | 634 | |||
XM_017029838.1 | XP_016885327.1 | 7 | 638 | |||
GenBank mRNA | AK298886.1 | - | 6 | 456 | ||
AK299194.1 | - | 7 | 519 | |||
AK310286.1 | - | 7 | 522 | |||
AK313544.1 | - | 7 | 516 | |||
AY438030.1 | - | 7 | 513 | |||
BC002999.1 | - | 7 | 515 | 2821917 | ||
BC006540.2 | - | 7 | 515 | 2821917 | ||
BU902387.1 | - | 653 | ||||
S68015.1 | - | 7 | 546 | |||
X64643.1 | - | 7 | 519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
