siRNA Details
Gene Symbol: TBC1D16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271844.1 | NP_001258773.1 | 4, 5 | 520 | ||
NM_001271845.1 | NP_001258774.1 | 4, 5 | 559 | |||
NM_001271846.1 | NP_001258775.1 | 4, 5 | 559 | |||
NM_019020.3 | NP_061893.2 | 8, 9 | 1675 | |||
XM_005257049.2 | XP_005257106.1 | 8, 9 | 1686 | |||
XM_005257050.3 | XP_005257107.1 | 8, 9 | 1689 | |||
XM_005257052.3 | XP_005257109.1 | 8, 9 | 1692 | |||
XM_006721694.3 | XP_006721757.1 | 8, 9 | 1692 | |||
XM_006721695.3 | XP_006721758.1 | 8, 9 | 1692 | |||
XM_011524322.2 | XP_011522624.1 | 8, 9 | 1650 | |||
XM_011524323.2 | XP_011522625.2 | 6, 7 | 1600 | |||
XM_011524324.2 | XP_011522626.1 | 4, 5 | 1144 | |||
XM_017024185.1 | XP_016879674.1 | 8, 9 | 1689 | |||
XM_017024186.1 | XP_016879675.1 | 6, 7 | 1427 | |||
XM_017024187.1 | XP_016879676.1 | 7, 8 | 1667 | |||
XM_017024188.1 | XP_016879677.1 | 6, 7 | 807 | |||
GenBank mRNA | AB449902.1 | - | 7, 8 | 1549 | ||
BC001525.2 | - | 4, 5 | 530 | |||
BC028290.1 | - | 8, 9 | 1665 | 4480654 | ||
BC036947.1 | - | 4, 5 | 564 | 5392579 | ||
CR936670.1 | - | 4, 5 | 525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
