siRNA Details
Gene Symbol: SIMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308195.1 | NP_001295124.1 | 7 | 2285 | ||
NM_001308196.1 | NP_001295125.1 | 9 | 2493 | |||
NM_001308200.1 | NP_001295129.1 | 4 | 741 | |||
NM_198567.5 | NP_940969.3 | 6 | 983 | |||
NR_131772.1 | 6 | 983 | ||||
XM_011534553.2 | XP_011532855.1 | 8 | 6290 | |||
XM_011534554.2 | XP_011532856.1 | 8 | 2162 | |||
XM_011534556.2 | XP_011532858.1 | 7 | 6159 | |||
XM_017009454.1 | XP_016864943.1 | 7 | 2093 | |||
XM_017009455.1 | XP_016864944.1 | 8 | 2403 | |||
XM_017009456.1 | XP_016864945.1 | 8 | 2299 | |||
GenBank mRNA | BC032390.1 | - | 4 | 709 | ||
BC037298.1 | - | 9 | 2494 | 4823991 | ||
BC073880.1 | - | 4 | 741 | |||
KF151170.1 | - | 7 | 2153 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
