siRNA Details
Gene Symbol: SIMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308195.1 | NP_001295124.1 | 6 | 2051 | ||
NM_001308196.1 | NP_001295125.1 | 8 | 2259 | |||
NM_001308200.1 | NP_001295129.1 | 3 | 507 | |||
NM_198567.5 | NP_940969.3 | 5 | 749 | |||
NR_131772.1 | 5 | 749 | ||||
XM_011534553.2 | XP_011532855.1 | 7 | 6056 | |||
XM_011534554.2 | XP_011532856.1 | 7 | 1928 | |||
XM_011534556.2 | XP_011532858.1 | 6 | 5925 | |||
XM_017009454.1 | XP_016864943.1 | 6 | 1859 | |||
XM_017009455.1 | XP_016864944.1 | 7 | 2169 | |||
XM_017009456.1 | XP_016864945.1 | 7 | 2065 | |||
GenBank mRNA | BC032390.1 | - | 3 | 475 | ||
BC037298.1 | - | 8 | 2260 | 4823991 | ||
BC066980.1 | - | 5 | 742 | 4821958 | ||
BC073880.1 | - | 3 | 507 | |||
KF151170.1 | - | 6 | 1919 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
