siRNA Details
Gene Symbol: CATSPERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021185.4 | NP_067008.3 | 11 | 1306 | ||
XM_005259114.4 | XP_005259171.1 | 11 | 1402 | |||
XM_005259115.4 | XP_005259172.1 | 11 | 1325 | |||
XM_006723308.3 | XP_006723371.1 | 11 | 1922 | |||
XM_006723310.2 | XP_006723373.1 | 11 | 1306 | |||
XM_006723311.2 | XP_006723374.1 | 11 | 1327 | |||
XM_011527174.2 | XP_011525476.1 | 11 | 1316 | |||
XM_011527175.2 | XP_011525477.1 | 11 | 1222 | |||
XM_011527176.2 | XP_011525478.1 | 11 | 1306 | |||
XM_011527177.2 | XP_011525479.1 | 11 | 1186 | |||
XM_011527178.2 | XP_011525480.1 | 12 | 1668 | |||
XM_011527179.1 | XP_011525481.1 | 11 | 1306 | |||
XM_011527182.2 | XP_011525484.1 | 10 | 1050 | |||
XM_017027054.1 | XP_016882543.1 | 9 | 919 | |||
GenBank mRNA | AK128220.1 | - | 11 | 1089 | ||
AK310460.1 | - | 7 | 636 | |||
AL133023.1 | - | 11 | 1922 | |||
BC094785.1 | - | 11 | 1226 | |||
BC109035.1 | - | 7 | 705 | 40028284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
