siRNA Details
Gene Symbol: SBSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166034.1 | NP_001159506.1 | 3 | 1805 | ||
NM_001166035.1 | NP_001159507.1 | 3 | 542 | |||
NM_198538.3 | NP_940940.1 | 4 | 776 | |||
XM_011526929.2 | XP_011525231.1 | 4 | 1771 | |||
XM_011526930.2 | XP_011525232.1 | 4 | 1727 | |||
XM_011526931.2 | XP_011525233.1 | 4 | 1717 | |||
GenBank mRNA | AA055367.1 | - | 3 | 329 | ||
AK291305.1 | - | 4 | 736 | |||
AY358701.1 | - | 4 | 776 | |||
BC063640.1 | - | 4 | 718 | 4750640 | ||
BX648076.1 | - | 3 | 1767 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
