siRNA Details
Gene Symbol: LINC00905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024335.2 | 2 | 441 | |||
NR_110321.1 | 2 | 151 | ||||
GenBank mRNA | AK058057.1 | - | 2 | 400 | ||
BC031284.1 | - | 2 | 151 | |||
BC069223.1 | - | 2 | 373 | |||
BC104221.2 | - | 1 | 338 | |||
DB461577.1 | - | 2 | 441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
