siRNA Details
Gene Symbol: MAP3K15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001671.3 | NP_001001671.3 | 17 | 2297 | ||
XM_011545507.2 | XP_011543809.2 | 18 | 2524 | |||
XM_011545508.2 | XP_011543810.2 | 18 | 2524 | |||
XM_011545510.2 | XP_011543812.1 | 11 | 3037 | |||
XM_011545511.2 | XP_011543813.1 | 8 | 1548 | |||
XM_017029511.1 | XP_016885000.1 | 18 | 2526 | |||
GenBank mRNA | AK131412.1 | - | 14 | 1625 | ||
AK131477.1 | - | 18 | 2357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
