siRNA Details
Gene Symbol: MAP3K15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001671.3 | NP_001001671.3 | 18, 19 | 2425 | ||
XM_011545507.2 | XP_011543809.2 | 19, 20 | 2652 | |||
XM_011545508.2 | XP_011543810.2 | 19, 20 | 2652 | |||
XM_011545510.2 | XP_011543812.1 | 12, 13 | 3165 | |||
XM_011545511.2 | XP_011543813.1 | 9, 10 | 1676 | |||
GenBank mRNA | AK131412.1 | - | 15, 16 | 1753 | ||
AK131477.1 | - | 19, 20 | 2485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
