siRNA Details
Gene Symbol: SLC45A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012509.3 | NP_001012527.1 | 4 | 1101 | ||
NM_001297417.2 | NP_001284346.2 | 3 | 775 | |||
NM_016180.4 | NP_057264.3 | 4 | 1101 | |||
GenBank mRNA | AF172849.1 | - | 4 | 1034 | ||
BC003597.1 | - | 3 | 744 | |||
BC064405.1 | - | 4 | 1067 | 5563039 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
