siRNA Details
Gene Symbol: SLC45A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012509.3 | NP_001012527.1 | 1 | 233 | ||
NM_001297417.2 | NP_001284346.2 | 1 | 233 | |||
NM_016180.4 | NP_057264.3 | 1 | 233 | |||
GenBank mRNA | AF172849.1 | - | 1 | 166 | ||
BC003597.1 | - | 1 | 202 | |||
BC064405.1 | - | 1 | 199 | 5563039 | ||
BQ677785.1 | - | 1 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
