siRNA Details
Gene Symbol: TBL1Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033284.1 | NP_150600.1 | 11 | 1410 | ||
NM_134258.1 | NP_599020.1 | 10 | 1379 | |||
NM_134259.1 | NP_599021.1 | 10 | 1315 | |||
XM_005262572.3 | XP_005262629.1 | 5 | 1074 | |||
XM_017030086.1 | XP_016885575.1 | 11 | 1399 | |||
XM_017030087.1 | XP_016885576.1 | 11 | 1399 | |||
GenBank mRNA | AF332220.1 | - | 11 | 1410 | ||
AF332221.1 | - | 10 | 1379 | |||
AF332222.1 | - | 10 | 1315 | |||
AK298210.1 | - | 11 | 1425 | |||
BC130471.1 | - | 7 | 893 | 40146502 | ||
BC130473.1 | - | 7 | 893 | 40146504 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
