siRNA Details
Gene Symbol: HELLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289067.1 | NP_001275996.1 | 22 | 2686 | ||
NM_001289068.1 | NP_001275997.1 | 21 | 2503 | |||
NM_001289069.1 | NP_001275998.1 | 21 | 2452 | |||
NM_001289070.1 | NP_001275999.1 | 19 | 2254 | |||
NM_001289071.1 | NP_001276000.1 | 22 | 2592 | |||
NM_001289072.1 | NP_001276001.1 | 19 | 2158 | |||
NM_001289073.1 | NP_001276002.1 | 20 | 2483 | |||
NM_001289074.1 | NP_001276003.1 | 22 | 2467 | |||
NM_001289075.1 | NP_001276004.1 | 20 | 2351 | |||
NM_018063.4 | NP_060533.2 | 21 | 2548 | |||
GenBank mRNA | AB074174.1 | - | 9 | 996 | ||
AB102716.1 | - | 22 | 2437 | |||
AB102717.1 | - | 20 | 2328 | |||
AB102718.1 | - | 21 | 2297 | |||
AB102719.1 | - | 19 | 2003 | |||
AB102720.1 | - | 20 | 2196 | |||
AB102721.1 | - | 19 | 2099 | |||
AB102722.1 | - | 22 | 2312 | |||
AB102723.1 | - | 22 | 2531 | |||
AB113248.1 | - | 21 | 2306 | |||
AB113249.1 | - | 20 | 2471 | |||
AF155827.1 | - | 21 | 2498 | |||
AK001201.1 | - | 7 | 706 | |||
AK314485.1 | - | 21 | 2537 | |||
AY007108.1 | - | 9 | 1005 | |||
BC015477.1 | - | 8 | 831 | |||
BC029381.1 | - | 9 | 936 | |||
BC030963.1 | - | 10 | 1151 | |||
BC031004.1 | - | 10 | 1151 | |||
BX538033.1 | - | 21 | 2487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
