siRNA Details
Gene Symbol: HELLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289067.1 | NP_001275996.1 | 22, 23 | 2711 | ||
NM_001289068.1 | NP_001275997.1 | 21, 22 | 2528 | |||
NM_001289069.1 | NP_001275998.1 | 21, 22 | 2477 | |||
NM_001289070.1 | NP_001275999.1 | 19, 20 | 2279 | |||
NM_001289071.1 | NP_001276000.1 | 22, 23 | 2617 | |||
NM_001289072.1 | NP_001276001.1 | 19, 20 | 2183 | |||
NM_001289073.1 | NP_001276002.1 | 20, 21 | 2508 | |||
NM_001289074.1 | NP_001276003.1 | 22, 23 | 2492 | |||
NM_001289075.1 | NP_001276004.1 | 20, 21 | 2376 | |||
NM_018063.4 | NP_060533.2 | 21, 22 | 2573 | |||
GenBank mRNA | AB074174.1 | - | 9, 10 | 1021 | ||
AB102716.1 | - | 22, 23 | 2462 | |||
AB102717.1 | - | 20, 21 | 2353 | |||
AB102718.1 | - | 21, 22 | 2322 | |||
AB102719.1 | - | 19, 20 | 2028 | |||
AB102720.1 | - | 20, 21 | 2221 | |||
AB102721.1 | - | 19, 20 | 2124 | |||
AB102722.1 | - | 22, 23 | 2337 | |||
AB102723.1 | - | 22, 23 | 2556 | |||
AB113248.1 | - | 21, 22 | 2331 | |||
AB113249.1 | - | 20, 21 | 2496 | |||
AF155827.1 | - | 21, 22 | 2523 | |||
AK001201.1 | - | 7, 8 | 731 | |||
AK314485.1 | - | 21, 22 | 2562 | |||
AY007108.1 | - | 9, 10 | 1030 | |||
BC015477.1 | - | 8, 9 | 856 | |||
BC029381.1 | - | 9, 10 | 961 | |||
BC030963.1 | - | 10, 11 | 1176 | |||
BC031004.1 | - | 10, 11 | 1176 | |||
BX538033.1 | - | 21, 22 | 2512 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
