siRNA Details
Gene Symbol: HELLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289067.1 | NP_001275996.1 | 21 | 2592 | ||
NM_001289068.1 | NP_001275997.1 | 20 | 2409 | |||
NM_001289069.1 | NP_001275998.1 | 20 | 2358 | |||
NM_001289070.1 | NP_001275999.1 | 18 | 2160 | |||
NM_001289071.1 | NP_001276000.1 | 21 | 2498 | |||
NM_001289072.1 | NP_001276001.1 | 18 | 2064 | |||
NM_001289073.1 | NP_001276002.1 | 19 | 2389 | |||
NM_001289074.1 | NP_001276003.1 | 21 | 2373 | |||
NM_001289075.1 | NP_001276004.1 | 19 | 2257 | |||
NM_018063.4 | NP_060533.2 | 20 | 2454 | |||
GenBank mRNA | AB074174.1 | - | 8 | 902 | ||
AB102716.1 | - | 21 | 2343 | |||
AB102717.1 | - | 19 | 2234 | |||
AB102718.1 | - | 20 | 2203 | |||
AB102719.1 | - | 18 | 1909 | |||
AB102720.1 | - | 19 | 2102 | |||
AB102721.1 | - | 18 | 2005 | |||
AB102722.1 | - | 21 | 2218 | |||
AB102723.1 | - | 21 | 2437 | |||
AB113248.1 | - | 20 | 2212 | |||
AB113249.1 | - | 20 | 2299 | |||
AF155827.1 | - | 20 | 2404 | |||
AK001201.1 | - | 6 | 612 | |||
AK314485.1 | - | 20 | 2443 | |||
AY007108.1 | - | 8 | 911 | |||
BC015477.1 | - | 7 | 737 | |||
BC029381.1 | - | 8 | 842 | |||
BC030963.1 | - | 9 | 1057 | |||
BC031004.1 | - | 9 | 1057 | |||
BX538033.1 | - | 20 | 2393 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
