siRNA Details
Gene Symbol: VPS39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301138.1 | NP_001288067.1 | 17 | 1837 | ||
NM_015289.3 | NP_056104.2 | 16 | 1804 | |||
XM_011521403.2 | XP_011519705.1 | 17 | 1806 | |||
XM_011521404.2 | XP_011519706.1 | 16 | 1809 | |||
XM_017022036.1 | XP_016877525.1 | 17 | 1805 | |||
XM_017022037.1 | XP_016877526.1 | 16 | 1809 | |||
XM_017022038.1 | XP_016877527.1 | 16 | 1992 | |||
XM_017022039.1 | XP_016877528.1 | 16 | 1803 | |||
GenBank mRNA | AB018313.2 | - | 16 | 1755 | ||
AF280814.1 | - | 17 | 1664 | |||
AF281052.1 | - | 16 | 1774 | |||
AK128403.1 | - | 1 | 12 | |||
BC015817.2 | - | 12 | 1364 | |||
BC068559.1 | - | 16 | 1773 | 5295166 | ||
BX537404.1 | - | 16 | 1783 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
