siRNA Details
Gene Symbol: VPS39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301138.1 | NP_001288067.1 | 24 | 2561 | ||
NM_015289.3 | NP_056104.2 | 23 | 2528 | |||
XM_011521403.2 | XP_011519705.1 | 24 | 2530 | |||
XM_011521404.2 | XP_011519706.1 | 23 | 2533 | |||
XM_017022036.1 | XP_016877525.1 | 24 | 2529 | |||
XM_017022037.1 | XP_016877526.1 | 23 | 2533 | |||
XM_017022038.1 | XP_016877527.1 | 23 | 2716 | |||
XM_017022039.1 | XP_016877528.1 | 23 | 2527 | |||
GenBank mRNA | AB018313.2 | - | 23 | 2479 | ||
AF280814.1 | - | 24 | 2388 | |||
AF281052.1 | - | 23 | 2498 | |||
AK025334.1 | - | 8 | 728 | |||
AK128403.1 | - | 6 | 1850 | |||
BC015817.2 | - | 19 | 2088 | |||
BC068559.1 | - | 23 | 2497 | 5295166 | ||
BX537404.1 | - | 23 | 2507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
